4,6-Dimethyl-2-mercaptopyrimidine ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Mercapto-4,6-dimethylpyrimidine
026733-25G
026733-100G
101822-756EA
29.23
USD
101822-756
101822-758
4,6-Dimethyl-2-mercaptopyrimidine ≥98%
4,6-Dimethyl-2-mercaptopyrimidine
Hazard: Irritant
Formula:
C₆H₈N₂S MW: 140.20 g/mol |
MDL Number:
MFCD00006079 CAS Number: 22325-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...