5-Methoxy-2-(trifluoromethyl)pyridine ≥98%
Supplier: Matrix Scientific
026731-1G
026731-5G
101822-748EA
70.77
USD
101822-748
101822-750
5-Methoxy-2-(trifluoromethyl)pyridine ≥98%
5-Methoxy-2-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₇H₆F₃NO MW: 177.13 g/mol |
MDL Number:
MFCD09864699 CAS Number: 216766-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...