2-Fluoro-m-toluidine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-3-methylaniline, 3-Amino-2-fluorotoluene

026299-1G 026299-5G 026299-25G
101822-030EA 20 USD
101822-030 101822-032 101822-034
2-Fluoro-m-toluidine ≥98%
2-Fluoro-m-toluidine

Hazard: Irritant

Formula: C₇H₈FN
MW: 125.14 g/mol
MDL Number: MFCD06410915
CAS Number: 1978-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR