2-Bromo-5-methyl-3-nitropyridine ≥98%
Supplier: Matrix Scientific
026294-5G
026294-25G
101822-008EA
27.69
USD
101822-008
101822-010
2-Bromo-5-methyl-3-nitropyridine ≥98%
2-Bromo-5-methyl-3-nitropyridine
Hazard: Irritant
Formula:
C₆H₅BrN₂O₂ MW: 217.0200 g/mol |
MDL Number:
MFCD08703667 CAS Number: 23056-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...