4-Cyano-7-azaindole
Supplier: Matrix Scientific
Synonyms:
4-Cyano-7-azaindole, 1H-Pyrrolo[2,3-b]pyridine-4-carbonitrile, 7-Azaindole-4-carbonitrile
026237-250MG
026237-1G
101821-890EA
1589.23
USD
101821-890
101821-892
4-Cyano-7-azaindole
4-Cyano-7-azaindole
Hazard: Irritant
Formula:
C₈H₅N₃ MW: 143.1500 g/mol |
MDL Number:
MFCD08272235 CAS Number: 344327-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...