1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 7-Azaindole-4-carboxylic acid

026235-250MG 026235-1G
101821-884EA 46.15 USD
101821-884 101821-886
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.148 g/mol
Melting Pt: >330
MDL Number: MFCD08272236
CAS Number: 479553-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR