Methyl-6-(hydroxymethyl)nicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: 6-(Hydroxymethyl)pyridine-3-carboxylate methyll ester, Methyl 6-(hydroxymethyl)nicotinate, 6-(Hydroxymethyl)nicotinic acid methyll ester, Methyl 6-(Hydroxymethyl)pyridine-3-carboxylate

026191-250MG 026191-1G
101821-794EA 807.69 USD
101821-794 101821-796
Methyl-6-(hydroxymethyl)nicotinate ≥98%
Methyl-6-(hydroxymethyl)nicotinate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD00643821
CAS Number: 56026-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR