5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-(trifluoromethyl)picolinate, Methyl 5-(trifluoromethyl)pyridine-2-carboxylate
026190-5G
026190-25G
101821-790EA
61.54
USD
101821-790
101821-792
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester ≥95%
5-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester
Hazard: Irritant
Formula:
C₈H₆F₃NO₂ MW: 205.136 g/mol |
MDL Number:
MFCD08741523 CAS Number: 124236-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...