2-Amino-4-chloro-3-iodopyridine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-iodopyridin-2-amine

026188-1G 026188-5G
101821-784EA 75.38 USD
101821-784 101821-786
2-Amino-4-chloro-3-iodopyridine
2-Amino-4-chloro-3-iodopyridine

Hazard: Irritant/Light Sensitive

Formula: C₅H₄ClIN₂
MW: 254.46 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD08741521
CAS Number: 417721-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR