2-Amino-5-bromonicotinonitrile

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-nicotinonitrile, 2-Amino-5-bromo-3-cyanopyridine

026175-250MG 026175-1G
101821-754EA 27.69 USD
101821-754 101821-756
2-Amino-5-bromonicotinonitrile
2-Amino-5-bromonicotinonitrile

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD08688591
CAS Number: 709652-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR