2-Amino-6-methoxypyridine ≥98%
Supplier: Matrix Scientific
026146-1G
026146-5G
101821-694EA
18.46
USD
101821-694
101821-696
2-Amino-6-methoxypyridine ≥98%
2-Amino-6-methoxypyridine
Hazard: Irritant
Formula:
C₆H₈N₂O MW: 124.14 g/mol |
MDL Number:
MFCD04972542 CAS Number: 17920-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...