Pivalamidine hydrochloride

Supplier: Matrix Scientific

020759-1G 020759-5G 020759-25G
101810-720EA 56.92 USD
101810-720 101810-722 101810-724
Pivalamidine hydrochloride
Pivalamidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂N₂
MDL Number: MFCD00051988
CAS Number: 18202-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR