Ethyl-2-aminothiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-aminothiophene-3-carboxylate

020677-5G 020677-25G
101810-572EA 98.46 USD
101810-572 101810-574
Ethyl-2-aminothiophene-3-carboxylate ≥95%
Ethyl-2-aminothiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD01566303
CAS Number: 31891-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR