5-Fluoro-2-iodobenzotrifluoride ≥97%
Supplier: Matrix Scientific
020598-5G
020598-25G
101810-402EA
26.15
USD
101810-402
101810-404
5-Fluoro-2-iodobenzotrifluoride ≥97%
5-Fluoro-2-iodobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₃F₄I |
MDL Number:
MFCD07369008 CAS Number: 41860-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...