Ethyl-5-methyl-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-methyl-1H-pyrrole-2-carboxylate

020594-1G 020594-5G
101810-390EA 27.69 USD
101810-390 101810-392
Ethyl-5-methyl-1H-pyrrole-2-carboxylate ≥97%
Ethyl-5-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₂
MW: 153.1800 g/mol
MDL Number: MFCD01006731
CAS Number: 3284-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR