6-Methoxynicotinonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methoxypyridine-5-carbonitrile

020587-1G 020587-5G
101810-372EA 26.15 USD
101810-372 101810-374
6-Methoxynicotinonitrile ≥97%
6-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.1400 g/mol
MDL Number: MFCD00084981
CAS Number: 15871-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR