3-Amino-5-methylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Amino-3-picoline
020581-5G
020581-25G
101810-346EA
21.54
USD
101810-346
101810-348
3-Amino-5-methylpyridine ≥97%
3-Amino-5-methylpyridine
Hazard: Irritant-Harmful
Formula:
C6H8N2 MW: 108.1411 g/mol |
MDL Number:
MFCD04112508 CAS Number: 3430-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...