3-Amino-5-fluoropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Fluoropyridin-3-amine

020572-1G 020572-5G
101810-318EA 15.38 USD
101810-318 101810-320
3-Amino-5-fluoropyridine ≥97%
3-Amino-5-fluoropyridine

Hazard: Irritant

Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD00234073
CAS Number: 210169-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR