3-Bromo-5-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

020565-5G 020565-25G 020565-100G
101810-294EA 26.15 USD
101810-294 101810-296 101810-298
3-Bromo-5-(trifluoromethyl)benzoic acid ≥98%
3-Bromo-5-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₄BrF₃O₂
MDL Number: MFCD03412186
CAS Number: 328-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR