1-Bromo-5-chloro-3-fluoro-2-iodobenzene ≥97%

Supplier: Matrix Scientific

020564-5G 020564-25G
101810-290EA 30.77 USD
101810-290 101810-292
1-Bromo-5-chloro-3-fluoro-2-iodobenzene ≥97%
1-Bromo-5-chloro-3-fluoro-2-iodobenzene

Hazard: Irritant

Formula: C₆H₂BrClFI
MDL Number: MFCD00142591
CAS Number: 201849-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR