4-Bromo-5-chloro-2-methylaniline ≥98%

Supplier: Matrix Scientific

020563-5G 020563-25G
101810-286EA 21.54 USD
101810-286 101810-288
4-Bromo-5-chloro-2-methylaniline ≥98%
4-Bromo-5-chloro-2-methylaniline

Hazard: Irritant

Formula: C₇H₇BrClN
MW: 220.5 g/mol
MDL Number: MFCD07782182
CAS Number: 30273-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR