2-Bromo-1-iodo-4-(trifluoromethoxy)benzene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(trifluoromethoxy)iodobenzene

020560-25G 020560-100G
101810-274EA 41.54 USD
101810-274 101810-276
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene ≥98%
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₃BrF₃IO
MW: 366.9018 g/mol
MDL Number: MFCD07369009
CAS Number: 883546-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR