2-Bromo-1,3-difluoro-5-iodobenzene ≥98%

Supplier: Matrix Scientific

020556-5G 020556-25G 020556-100G
101810-256EA 84.62 USD
101810-256 101810-258 101810-260
2-Bromo-1,3-difluoro-5-iodobenzene ≥98%
2-Bromo-1,3-difluoro-5-iodobenzene

Hazard: Irritant

Formula: C₆H₂BrF₂I
MDL Number: MFCD06797752
CAS Number: 155906-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR