2-Bromo-1-chloro-4-trifluoromethoxybenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-chloro-4-(trifluoromethoxy)benzene

020555-5G 020555-25G
101810-252EA 29.23 USD
101810-252 101810-254
2-Bromo-1-chloro-4-trifluoromethoxybenzene ≥98%
2-Bromo-1-chloro-4-trifluoromethoxybenzene

Hazard: Irritant

Formula: C₇H₃BrClF₃O
MW: 275.46 g/mol
MDL Number: MFCD06797751
CAS Number: 468075-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR