L-Tyrosine methyl ester

Supplier: Matrix Scientific

020549-5G 020549-25G
101810-226EA 63.08 USD
101810-226 101810-228
L-Tyrosine methyl ester
L-Tyrosine methyl ester

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00002392
CAS Number: 1080-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR