(2-Methoxyphenyl)hydrazine

Supplier: Matrix Scientific

020477-500MG 020477-2.5G
101810-092EA 484.62 USD
101810-092 101810-094
(2-Methoxyphenyl)hydrazine
(2-Methoxyphenyl)hydrazine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00460677
CAS Number: 18312-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR