3-Oxo-3-pyridin-3-ylpropionic acid ethyl ester

Supplier: Matrix Scientific

020471-1G 020471-5G
101810-078EA 127.69 USD
101810-078 101810-080
3-Oxo-3-pyridin-3-ylpropionic acid ethyl ester
3-Oxo-3-pyridin-3-ylpropionic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD00160418
CAS Number: 6283-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR