7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Supplier: Matrix Scientific

020442-500MG 020442-1G
101810-022EA 495.38 USD
101810-022 101810-024
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₈N₄O₂
MW: 192.18 g/mol
MDL Number: MFCD03011802
CAS Number: 2627-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR