Methyl-3-(chlorosulfonyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(chlorosulphonyl)benzoate

020428-1G 020428-5G
101809-996EA 75.38 USD
101809-996 101809-998
Methyl-3-(chlorosulfonyl)benzoate ≥95%
Methyl 3-(chlorosulphonyl)benzoate

Hazard: Irritant

Formula: C₈H₇ClO₄S
MW: 234.6600 g/mol
MDL Number: MFCD06408800
CAS Number: 63555-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR