1-Difluoromethyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

020332-500MG 020332-1G
101809-830EA 1627.69 USD
101809-830 102663-620
1-Difluoromethyl-3-methyl-1H-pyrazole-4-sulfonyl chloride
1-Difluoromethyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₅H₅ClF₂N₂O₂S
MDL Number: MFCD04970130
CAS Number: 957490-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR