3-Fluoro-4-methoxybenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methoxybenzene-1-sulphonyl chloride

020070-1G 020070-5G
101809-342EA 35.38 USD
101809-342 101809-344
3-Fluoro-4-methoxybenzenesulfonyl chloride ≥95%
3-Fluoro-4-methoxybenzenesulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₆ClFO₃S
Melting Pt: 77…80 °C
MDL Number: MFCD02091380
CAS Number: 67475-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR