4-Methoxy-2,3,6-trimethylbenzaldehyde

Supplier: Matrix Scientific

020036-1G 020036-5G
101809-292EA 138.46 USD
101809-292 101809-294
4-Methoxy-2,3,6-trimethylbenzaldehyde
4-Methoxy-2,3,6-trimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00456729
CAS Number: 54344-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR