2-Amino-6-chlorophenol
Supplier: Matrix Scientific
020009-1G
020009-5G
101809-222EA
35.38
USD
101809-222
101809-224
2-Amino-6-chlorophenol
2-Amino-6-chlorophenol
Hazard: Toxic/Stored under Argon/Light Sensitive/Air Sensitive
Formula:
C₆H₆ClNO MW: 143.5700 g/mol Melting Pt: 80-81°C Flash Pt: 103°C |
MDL Number:
MFCD07801210 CAS Number: 38191-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...