4-Amino-4'-methyldiphenyl ether

Supplier: Matrix Scientific

Synonyms: 4-(p-Tolyloxy)aniline, 4-Aminophenyl p-Tolyl Ether 4-(p-Tolyloxy)aniline, 4-Amino-4'-methyldiphenyl Ether, 4-Aminophenyl p-Tolyl Ether

022471-5G 022471-25G
101814-100EA 235.38 USD
101814-100 101814-102
4-Amino-4'-methyldiphenyl ether
4-Amino-4'-methyldiphenyl ether

Hazard: Irritant

Formula: C₁₃H₁₃NO
MDL Number: MFCD00059858
CAS Number: 41295-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR