1-Methyl-1H-indole-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Indole-2-carboxylic acid, 1-methyl-, 1-Methylindole-2-carboxylic acid

016213-1G 016213-5G
101804-314EA 21.54 USD
101804-314 101804-316
1-Methyl-1H-indole-2-carboxylic acid ≥98%
1-Methyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00005801
CAS Number: 16136-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR