(4-Methoxybenzyl)hydrazine dihydrochloride ≥95%

Supplier: Matrix Scientific

016133-1G 016133-5G
101804-166EA 67.69 USD
101804-166 101804-168
(4-Methoxybenzyl)hydrazine dihydrochloride ≥95%
(4-Methoxybenzyl)hydrazine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₄Cl₂N₂O
Melting Pt: 194…195 °C (dec.)
MDL Number: MFCD06800897
CAS Number: 412327-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR