(3,4-Difluorophenyl)hydrazine hydrochloride
Supplier: Matrix Scientific
Synonyms:
(3,4-Difluorophenyl)hydrazine xhydrochloride, 3,4-Difluorophenylhydrazinium chloride, 1-(3,4-Difluorophenyl)hydrazine hydrochloride
016122-5G
016122-25G
101804-144EA
116.92
USD
101804-144
101804-146
(3,4-Difluorophenyl)hydrazine hydrochloride
(3,4-Difluorophenyl)hydrazine hydrochloride
Hazard: Irritant
Formula:
C₆H₇ClF₂N₂ MW: 180.59 g/mol |
MDL Number:
MFCD03094170 CAS Number: 875664-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...