N-Methyltetrahydro-2H-pyran-4-amine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Methylamino)tetrahydro-2H-pyran

016105-250MG 016105-1G
101804-102EA 32.31 USD
101804-102 101804-104
N-Methyltetrahydro-2H-pyran-4-amine ≥98%
N-Methyltetrahydro-2H-pyran-4-amine

Hazard: Irritant

Formula: C₆H₁₃NO
MW: 115.1700 g/mol
MDL Number: MFCD06658492
CAS Number: 220641-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR