3,5-Dichloropicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloropyridine-2-carboxylic acid

016030-5G 016030-25G
101803-932EA 32.31 USD
101803-932 101803-934
3,5-Dichloropicolinic acid ≥95%
3,5-Dichloropicolinic acid

Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192.0000 g/mol
Melting Pt: 153…154 °C
MDL Number: MFCD03230005
CAS Number: 81719-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR