2-Aminoisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid

016027-5G 016027-25G
101803-920EA 61.54 USD
101803-920 101803-922
2-Aminoisonicotinic acid ≥98%
2-Aminoisonicotinic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 300-350°C (dec)
MDL Number: MFCD00233714
CAS Number: 13362-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR