5-Chloro-2-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

015723-5G 015723-25G
101803-882EA 92.31 USD
101803-882 101803-884
5-Chloro-2-methoxybenzaldehyde ≥95%
5-Chloro-2-methoxybenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₈H₇ClO₂
MW: 170.60 g/mol
Boiling Pt: 152°C/15 Torr
Melting Pt: 75-79°C
MDL Number: MFCD00033854
CAS Number: 7035-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR