4-Methoxy-3,5-dimethylbenzaldehyde
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethyl-4-methoxybenzaldehyde
015659-1G
015659-5G
101803-868EA
75.38
USD
101803-868
101803-870
4-Methoxy-3,5-dimethylbenzaldehyde
4-Methoxy-3,5-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD02261755 CAS Number: 39250-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...