5-Nitro-2-(trifluoromethoxy)aniline ≥98%
Supplier: Matrix Scientific
012904-5G
012904-25G
101803-596EA
44.62
USD
101803-596
101803-598
5-Nitro-2-(trifluoromethoxy)aniline ≥98%
5-Nitro-2-(trifluoromethoxy)aniline
Hazard: Irritant
Formula:
C₇H₅F₃N₂O₃ MW: 222.12 g/mol |
MDL Number:
MFCD06248592 CAS Number: 158579-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...