3-Perfluorohexyl-5-trifluoromethylpyrazole ≥97%

Supplier: Matrix Scientific

012876-1G 012876-5G
101803-556EA 56.92 USD
101803-556 101803-558
3-Perfluorohexyl-5-trifluoromethylpyrazole ≥97%
3-Perfluorohexyl-5-trifluoromethylpyrazole

Hazard: Irritant

Formula: C₁₀H₂F₁₆N₂
MDL Number: MFCD06248656
CAS Number: 1240309-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR