5,5,6,6,6-Pentafluorohexane-2,4-dione ≥97%
Supplier: Matrix Scientific
012863-25G
012863-100G
101803-534EA
158.46
USD
101803-534
101803-536
5,5,6,6,6-Pentafluorohexane-2,4-dione ≥97%
5,5,6,6,6-Pentafluorohexane-2,4-dione
Hazard: Irritant
Formula:
C₆H₅F₅O₂ Boiling Pt: 111…112 °C (631 Torr) |
MDL Number:
MFCD00466808 CAS Number: 356-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...