6-Iodo-1,1,1,2,2-pentafluorohexane ≥97%
Supplier: Matrix Scientific
012857-1G
012857-5G
101803-520EA
115.38
USD
101803-520
101803-522
6-Iodo-1,1,1,2,2-pentafluorohexane ≥97%
6-Iodo-1,1,1,2,2-pentafluorohexane
Hazard: Irritant
Formula:
C₆H₈F₅I Boiling Pt: 62-64°C/15 Torr |
MDL Number:
MFCD06247809 CAS Number: 344452-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...