2,6-Dichloro-5-fluoronicotinonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloro-3-cyano-5-fluoropyridine, 2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile

012670-25G 012670-100G
101803-164EA 93.85 USD
101803-164 101803-166
2,6-Dichloro-5-fluoronicotinonitrile ≥98%
2,6-Dichloro-5-fluoronicotinonitrile

Hazard: Irritant

Formula: C₆HCl₂FN₂
MW: 190.99 g/mol
Melting Pt: 91-93°C
MDL Number: MFCD01863289
CAS Number: 82671-02-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR