2,6-Dichloro-3-cyano-4-picoline ≥98%
Supplier: Matrix Scientific
012641-25G
012641-100G
101803-070EA
20
USD
101803-070
101803-072
2,6-Dichloro-3-cyano-4-picoline ≥98%
2,6-Dichloro-3-cyano-4-picoline
Hazard: Irritant
Formula:
C₇H₄Cl₂N₂ Boiling Pt: 116…118 °C (5 Torr) Melting Pt: 110 °C |
MDL Number:
MFCD00052633 CAS Number: 875-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...