1,1,1,2-Tetrafluoro-2-(trifluoromethyl)-6-iodohexane

Supplier: Matrix Scientific

012640-5G 012640-25G
101803-066EA 135.38 USD
101803-066 101803-068
1,1,1,2-Tetrafluoro-2-(trifluoromethyl)-6-iodohexane
1,1,1,2-Tetrafluoro-2-(trifluoromethyl)-6-iodohexane

Hazard: Irritant

Formula: C₇H₈F₇I
MDL Number: MFCD06248783
CAS Number: 99324-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR