2,6-Dihydroxy-3,4-dimethylpyridine ≥98%
Supplier: Matrix Scientific
012632-5G
012632-25G
101803-030EA
566.15
USD
101803-030
101803-032
2,6-Dihydroxy-3,4-dimethylpyridine ≥98%
2,6-Dihydroxy-3,4-dimethylpyridine
Hazard: Irritant
Formula:
C₇H₉NO₂ Melting Pt: 187 °C |
MDL Number:
MFCD05662686 CAS Number: 84540-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...